3-Hydroxypiperidine hydrochloride 99%

Warning

Synonyms: 3-Hydroxypiperidine Hydrochloride

AC17432-1000 AC17432-0050 AC17432-0250
103510-984EA 1283.7 USD
103510-984 103509-248 103509-246
3-Hydroxypiperidine hydrochloride 99%
3-Hydroxypiperidine hydrochloride
Formula: C₅H₁₂ClNO
MW: 137.61 g/mol
Melting Pt: 196.6…198.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00012772
CAS Number: 64051-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR