2-Iodo-3-pyridinol 97%
Warning
Synonyms: 3-Hydroxy-2-iodopyridine
AC43639-0010 AC43639-0050
103515-800EA 152.7 USD
103515-800 103515-798
2-Iodo-3-pyridinol 97%
2-Iodo-3-pyridinol
Order Now
Specification Test Results
Appearance White to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Elemental Analysis 26.08 to 28.25 % C
Elemental Analysis 6.09 to 6.59 % N