3-Hydroxy-2,4,6-triiodobenzoic acid 97%

Danger

AC45324-0250 AC45324-0050 AC45324-1000
103517-404EA 384 USD
103517-404 103517-402 103517-400
3-Hydroxy-2,4,6-triiodobenzoic acid 97%
3-Hydroxy-2,4,6-triiodobenzoic acid
Formula: C₇H₃I₃O₃
Melting Pt: 172.5…174.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00020305
CAS Number: 53279-72-4

Order Now

Specification Test Results

Appearance Off-white to beige to light brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR