3-Guanidinopropionic acid 97%

Warning

Synonyms: β-Guanidinopropionic acid3-Guanidinopropanoic acid

AC42666-0010 AC42666-0050
103512-620EA 55.6 USD
103512-620 103512-622
3-Guanidinopropionic acid 97%
3-Guanidinopropionic acid
Formula: C₄H₉N₃O₂
MW: 131.13 g/mol
Melting Pt: 222 °C
Storage Temperature: Ambient
MDL Number: MFCD00045939
CAS Number: 353-09-3

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Melting point 208 to 212 °C
NMR (1H-NMR) Consistent with structure
Titration with HClO4 ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR