3-Fluoro-4-methoxyphenylboronic acid ≥97%

Warning

Synonyms: 3-Fluoro-4-methoxybenzeneboronic acid

AC39394-0010
103514-406EA 39.2 USD
103514-406
3-Fluoro-4-methoxyphenylboronic acid ≥97%
3-Fluoro-4-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 211…213 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00807404
CAS Number: 149507-26-6

Order Now

Specification Test Results

Appearance White to tan Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR