3-Fluoro-2-formylpyridine 97%

Warning

Synonyms: 3-Fluoropicolinaldehyde3-Fluoro-2-pyridinecarboxaldehyde3-Fluoropyridine-2-carboxaldehyde

AC44862-0050 AC44862-0010
103516-838EA 66.7 USD
103516-838 103516-840
3-Fluoro-2-formylpyridine 97%
3-Fluoro-2-formylpyridine
Formula: C₆H₄FNO
MW: 125.1 g/mol
Melting Pt: 51…54 °C
Storage Temperature: Ambient
MDL Number: MFCD07781234
CAS Number: 31224-43-8

Order Now

Specification Test Results

Appearance Light yellow to brown Low melting solid
Infrared spectrum Conforms
Melting point 48 to 53 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR