3-Fluoro-2-nitropyridine 98%

Warning

AC45245-0010 AC45245-0050
103517-316EA 556 USD
103517-316 103517-314
3-Fluoro-2-nitropyridine 98%
3-Fluoro-2-nitropyridine
Formula: C₅H₃FN₂O₂
Melting Pt: 33…37 °C
Storage Temperature: Ambient
MDL Number: MFCD04114127
CAS Number: 54231-35-5

Order Now

Specification Test Results

Appearance Yellow Crystalline solid
Infrared spectrum Conforms
Melting point 33 to 37 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR