3-Cyano-5-fluorophenylboronic acid 97%

Warning

Synonyms: (3-Cyano-5-fluorophenyl)boronic acid

AC43046-0010
103515-278EA 262 USD
103515-278
3-Cyano-5-fluorophenylboronic acid 97%
3-Cyano-5-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
MW: 164.9300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09263608
CAS Number: 304858-67-1

Order Now

Specification Test Results

Appearance White Crystalline solid
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration Argentometric 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR