3-Chlorophenyl acetylene 97%
|
Warning
|
Synonyms:
(3-Chlorophenyl)acetylene
, 1-Chloro-3-ethynylbenzene
, 3-Chlorophenylacetylene
AC43313-0050
AC43313-0010
103515-490EA
195
USD
103515-490
103515-488
3-Chlorophenyl acetylene 97%
3-Chlorophenyl acetylene
Formula:
C₈H₅Cl MW: 136.578 g/mol Boiling Pt: 178…179 °C (760 mmHg) Flash Pt: 63 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03839984 CAS Number: 766-83-6 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...