3-Chloro-4-methoxybenzaldehyde 97%

Warning

AC43531-0010 AC43531-0050
103465-348EA 51.6 USD
103465-348 103465-350
3-Chloro-4-methoxybenzaldehyde 97%
3-Chloro-4-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
Boiling Pt: 128 °C (5 mmHg)
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00016601
CAS Number: 4903-09-7

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 54 to 58 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR