3-Chloro-2-methoxy-4-pyridineboronic acid 95%

Warning

Synonyms: 3-Chloro-2-methoxypyridine-4-boronic acid

AC44877-0050 AC44877-0010 AC44877-2500
103517-034EA 806.4 USD
103517-034 103517-032 103517-036
3-Chloro-2-methoxy-4-pyridineboronic acid 95%
3-Chloro-2-methoxy-4-pyridineboronic acid
Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
Melting Pt: 140 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06798255
CAS Number: 957060-88-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 137 to 142 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR