3-Buten-1-amine 97%
|
Danger
|
Synonyms:
3-Butenylamine
, 4-Amino-1-butene
AC45505-0050
AC45505-0010
103529-052EA
168
USD
103529-052
103529-054
3-Buten-1-amine 97%
3-Buten-1-amine
Formula:
C₄H₉N MW: 71.12 Boiling Pt: 83 °C (760 mmHg) Flash Pt: –5 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425859 CAS Number: 2524-49-4 UN: 2733 ADR: 3,II |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4215 to 1.4275 (20 °C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...