3-Bromothiophene-2-carbaldehyde 97%

Warning

Synonyms: 3-Bromothiophene-2-carboxaldehyde

AC45677-0050 AC45677-0010 AC45677-0250
103518-414EA 38.6 USD
103518-414 103518-416 103518-418
3-Bromothiophene-2-carbaldehyde 97%
3-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
MW: 191.05 g/mol
Boiling Pt: 92 °C (5 mmHg)
Melting Pt: 24…25 °C
Density: 1.81 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00126680
CAS Number: 930-96-1

Order Now

Specification Test Results

Appearance Yellow to brown Liquid or low melting solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR