3-Bromo-6-fluoroimidazo[1,2-a]pyridine 97%

Warning

AC45347-0010 AC45347-0050
103463-654EA 267.9 USD
103463-654 103463-656
3-Bromo-6-fluoroimidazo[1,2-a]pyridine 97%
3-Bromo-6-fluoroimidazo[1,2-a]pyridine
Formula: C₇H₄BrFN₂
Melting Pt: 138…141 °C
Storage Temperature: Ambient
MDL Number: MFCD12827770
CAS Number: 1186405-11-7

Order Now

Specification Test Results

Appearance Off-white to light brown powder
Infrared spectrum Conforms
Melting point 133 to 141 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR