3-Aminocoumarin 97%

Warning

Synonyms: 3-Amino-2H-chromen-2-one

AC44951-0010
103465-262EA 217.1 USD
103465-262
3-Aminocoumarin 97%
3-Aminocoumarin
Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 135…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00016965
CAS Number: 1635-31-0

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 135 to 139 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR