3-Amino-5-ethyl-1H-pyrazole 97%

Warning

Synonyms: 3-Amino-5-ethylpyrazole

AC44671-0050 AC44671-0010
103516-660EA 148 USD
103516-660 103516-818
3-Amino-5-ethyl-1H-pyrazole 97%
3-Amino-5-ethyl-1H-pyrazole
Formula: C₅H₉N₃
MW: 111.15 g/mol
Storage Temperature: Freezer
MDL Number: MFCD08060982
CAS Number: 1904-24-1

Order Now

Specification Test Results

Appearance Yellow to orange to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR