3-Bromo-5-(trifluoromethyl)aniline 97%

Warning

Synonyms: 3-Amino-5-bromobenzotrifluoride5-Bromo-α,α,α-trifluoro-m-toluidine3-Amino-5-bromo-α,α,α-trifluorotoluene

AC44266-0250 AC44266-0050
103465-234EA 75.3 USD
103465-234 103465-236
3-Bromo-5-(trifluoromethyl)aniline 97%
3-Bromo-5-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 220…223 °C (760 mmHg)
Density: 1.697 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00236205
CAS Number: 54962-75-3

Order Now

Specification Test Results

Appearance Colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR