3-Amino-2-cyclohexen-1-one 95%

Warning

AC21931-0050 AC21931-0250
103465-210EA 119.4 USD
103465-210 103465-212
3-Amino-2-cyclohexen-1-one 95%
3-Amino-2-cyclohexen-1-one
Formula: C₆H₉NO
MW: 111.14 g/mol
Melting Pt: 133…135 °C
Storage Temperature: Ambient
MDL Number: MFCD00013783
CAS Number: 5220-49-5

Order Now

Specification Test Results

Appearance Yellow to tan Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 127 to 134 °C
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR