3-Acetylphenanthrene 97%

Warning

Synonyms: 1-(phenanthren-3-yl)ethanone1-(phenanthren-3-yl)ethan-1-oneEthanone, 1-(3-phenanthrenyl)-

AC10264-0250
103465-188EA 392 USD
103465-188
3-Acetylphenanthrene 97%
3-Acetylphenanthrene
Formula: C₁₆H₁₂O
MW: 220.27 g/mol
Melting Pt: 73…76 °C
Storage Temperature: Ambient
MDL Number: MFCD00001169
CAS Number: 2039-76-1

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 71 to 76 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR