3-[(Benzyloxycarbonyl)amino]propionaldehyde 95%

Warning

Synonyms: N-Phenylmethoxycarbonyl-3-aminopropanal3-[(Benzyloxycarbonyl)amino]-1-propanalBenzyl 3-oxopropylcarbamate

AC42772-0010
103514-998EA 71.4 USD
103514-998
3-[(Benzyloxycarbonyl)amino]propionaldehyde 95%
3-[(Benzyloxycarbonyl)amino]propionaldehyde
Formula: C₁₁H₁₃NO₃
MW: 207.229 g/mol
Melting Pt: 52…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664298
CAS Number: 65564-05-8

Order Now

Specification Test Results

Appearance White to yellow Powder or solid
Infrared spectrum Conforms
Melting point 52 to 61 °C
GC ≥94 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR