3,5-Dichloroisonicotinic acid 98%

Warning

Synonyms: 3,5-Dichloropyridine-4-carboxylic acid

AC39902-0010 AC39902-0050
103465-128EA 260.7 USD
103465-128 103465-126
3,5-Dichloroisonicotinic acid 98%
3,5-Dichloroisonicotinic acid
Formula: C₆H₃Cl₂NO₂
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD01861975
CAS Number: 13958-93-5

Order Now

Specification Test Results

Appearance White to brown Powder or crystals
Infrared spectrum Conforms
Titration Acid ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR