3,5-Dibromosalicylic acid 97%

Warning

Synonyms: 3,5-Dibromo-2-hydroxybenzoic acid

AC11292-0250
103509-068EA 52 USD
103509-068
3,5-Dibromosalicylic acid 97%
3,5-Dibromosalicylic acid
Formula: C₇H₄Br₂O₃
MW: 295.91 g/mol
Melting Pt: 225…228 °C
Storage Temperature: Ambient
MDL Number: MFCD00002441
CAS Number: 3147-55-5
EINECS: 221-570-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 225 to 229 °C
HPLC ≥96.0 %
Residue after evaporation ≤0.002 %
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤10 ppm
Copper (Cu) ≤5 ppm
Reducing substances Passes test
Ammonia (NH3) ≤20 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR