3,3-Dimethylbutanone 96%

Danger

Synonyms: tert-Butyl methyl ketonePinacolone1,1,1-Trimethylacetone3,3-Dimethyl-2-butanoneMethyl tert-butyl ketone

AC13125-5000 AC13125-1000 AC13125-0025
103525-402EA 273 USD
103525-402 103525-398 103525-400
3,3-Dimethylbutanone 96%
3,3-Dimethylbutanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 106 °C (760 mmHg)
Melting Pt: –52.5 °C
Density: 0.8 g/cm³
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008846
CAS Number: 75-97-8
EINECS: 200-920-4
UN: 1224
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3950 to 1.3980 (20 °C, 589 nm)
GC ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR