3,3-Dimethoxypropiononitrile 90%

Warning

Synonyms: 3,3-Dimethoxypropanenitrile

AC44626-0250 AC44626-1000
103465-036EA 150 USD
103465-036 103465-038
3,3-Dimethoxypropiononitrile 90%
3,3-Dimethoxypropiononitrile
Formula: C₅H₉NO₂
MW: 115.13 g/mol
Boiling Pt: 195 °C (760 mmHg)
Density: 1.026 g/cm³
Flash Pt: 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00044797
CAS Number: 57597-62-3

Order Now

Specification Test Results

Appearance Colorless to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4120 to 1.4160 (20 °C, 589 nm)
GC ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR