3,3',5-Triiodo-L-thyronine 95%

Warning

Synonyms: (S)-2-Amino-3-(4-(4-hydroxy-3-iodophenoxy)-3,5-diiodophenyl)propanoic acid

AC43726-0050 AC43726-0010
103515-858EA 280 USD
103515-858 103515-860
3,3',5-Triiodo-L-thyronine 95%
3,3',5-Triiodo-L-thyronine
Formula: C₁₅H₁₂I₃NO₄
MW: 650.98 g/mol
Melting Pt: 234…238 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002593
CAS Number: 6893-02-3

Order Now

Specification Test Results

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥94 %
Specific optical rotation +18° to +28° (20 °C, 589 nm) (c=2,ethanol/0.5N HCl(4:1))

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR