3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%
|
Warning
|
Synonyms:
3,3',5,5'-Tetra-tert-butyldiphenoquinone
, 3,3',5,5'-Tetra-tert-butyl-[1,1'-bi(cyclohexylidene)]-2,2',5,5'-tetraene-4,4'-dione
AC44830-0020
AC44830-0100
103465-024EA
107
USD
103465-024
103465-026
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone 98%
3,3',5,5'-Tetra-tert-butyl-4,4'-diphenoquinone
Formula:
C₂₈H₄₀O₂ MW: 408.63 g/mol Melting Pt: 247…248 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051798 CAS Number: 2455-14-3 |
Specification Test Results
Appearance | Yellow to brown to red Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...