3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98%

Warning

Synonyms: Diethyl 3,4-Dihydro-4-oxo-1,2,3-benzotriazin-3-yl PhosphateDiethyl (4-oxobenzo[d][1,2,3]triazin-3(4H)-yl) phosphateDEPBT

AC44153-0050
103513-456EA 55.5 USD
103513-456
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one 98%
3-(Diethoxyphosphoryloxy)-1,2,3-benzotriazin-4(3H)-one
Formula: C₁₁H₁₄N₃O₅P
MW: 299.22 g/mol
Melting Pt: 73…77 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01236967
CAS Number: 165534-43-0

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR