2-Piperidinemethanol 97%

Warning

Synonyms: 2-(Hydroxymethyl)piperidine

AC16803-0050 AC16803-0250 AC16803-1000
103464-910EA 474 USD
103464-910 103464-908 103464-906
2-Piperidinemethanol 97%
2-Piperidinemethanol
Formula: C₆H₁₃NO
MW: 115.18 g/mol
Boiling Pt: 100…102 °C (8hPa)
Melting Pt: 58…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00005987
CAS Number: 3433-37-2
EINECS: 222-333-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR