(±)-2-Phenyl-3-butyn-2-ol 98%

Warning

Synonyms: 2-Phenylbut-3-yn-2-ol

AC19969-0050 AC19969-0250 AC19969-1000
103464-868EA 44.5 USD
103464-868 103464-870 103464-872
(±)-2-Phenyl-3-butyn-2-ol 98%
(±)-2-Phenyl-3-butyn-2-ol
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 102…103 °C (12 mmHg)
Melting Pt: 50…52 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00004454
CAS Number: 127-66-2
EINECS: 204-855-2

Order Now

Specification Test Results

Appearance Semitransparent white Crystals
Infrared spectrum Conforms
Melting point 46 to 52 °C
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR