2-Phenoxyphenylboronic acid 95%

Warning

Synonyms: 2-Phenoxybenzeneboronic acid

AC44402-0100
103516-302EA 140 USD
103516-302
2-Phenoxyphenylboronic acid 95%
2-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01001592
CAS Number: 108238-09-1

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR