2-Naphthol 98%

Warning

Synonyms: β-Naphthol2-Hydroxynaphthaleneβ-Hydroxynaphthalene

AC15697-2500 AC15697-0010 AC15697-0050
103525-904EA 228 USD
103525-904 103525-906 103525-902
2-Naphthol 98%
2-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 285…286 °C (760 mmHg)
Melting Pt: 120…124 °C
Flash Pt: 153 °C
Storage Temperature: Ambient
MDL Number: MFCD00004067
CAS Number: 135-19-3
EINECS: 205-182-7
UN: 3077
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR