2-Methyl-1,3-dithiane 97%

AC44326-0010 AC44326-0050
103464-758EA 63.6 USD
103464-758 103464-756
2-Methyl-1,3-dithiane 97%
2-Methyl-1,3-dithiane
Formula: C₅H₁₀S₂
Boiling Pt: 66 °C (5Torr)
Density: 1.12 g/cm³
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00014652
CAS Number: 6007-26-7

Order Now

Specification Test Results

Appearance Colorless to brown Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR