Ethylene glycol monomethyl ether (2-methoxyethanol) 99.5+% for analysis

Danger

Synonyms: Ethylene glycol monomethyl ether

AC14936-0025 AC14936-0010
103525-710EA 79.05 USD
103525-710 103525-712
Ethylene glycol monomethyl ether (2-methoxyethanol) 99.5+% for analysis
Ethylene glycol monomethyl ether (2-Methoxyethanol)
Formula: C₃H₈O₂
MW: 76.1 g/mol
Boiling Pt: 124 °C (760 mmHg)
Melting Pt: –85 °C
Density: 0.96 g/cm³
Flash Pt: 38 °C
Storage Temperature: Ambient
MDL Number: MFCD00002867
CAS Number: 109-86-4
EINECS: 203-713-7
UN: 1188
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR