2-Methoxy-3-nitropyridine 97%

Warning

AC45487-0250 AC45487-0100
103463-560EA 163.7 USD
103463-560 103463-558
2-Methoxy-3-nitropyridine 97%
2-Methoxy-3-nitropyridine
Formula: C₆H₆N₂O₃
Melting Pt: 50…53 °C
Storage Temperature: Ambient
MDL Number: MFCD00023459
CAS Number: 20265-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR