2-Mercaptoethanesulfonic acid sodium salt (mesna) 98% analytical standard

Warning

Synonyms: Sodium 2-mercaptoethanesulfonateSodium 2-mercaptoethanesulphonate

AC44315-0250 AC44315-0050 AC44315-0100
103516-282EA 180 USD
103516-282 103516-280 103516-278
2-Mercaptoethanesulfonic acid sodium salt (mesna) 98% analytical standard
2-Mercaptoethanesulphonic acid sodium salt (Mesna)
Formula: C₂H₅NaO₃S₂
MW: 164.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007535
CAS Number: 19767-45-4

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Titration Iodometric ≥97.5 to 102 % (On dried substance)
Loss on drying ≤2 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR