2-Imidazolidinone, anhydrous 96%

Warning

Synonyms: 2-Imidazolidone

AC41227-0250 AC41227-0010 AC41227-2500
103463-532EA 24.2 USD
103463-532 103463-534 103463-530
2-Imidazolidinone, anhydrous 96%
2-Imidazolidinone
Formula: C₃H₆N₂O
MW: 86.0925 g/mol
Boiling Pt: 187 °C (10Torr)
Melting Pt: 131.4…132.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00005257
CAS Number: 120-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR