5-Methylsalicylaldehyde 99%

Warning

Synonyms: 2-Hydroxy-5-methylbenzaldehyde

AC39263-0010 AC39263-0050
103464-702EA 30.2 USD
103464-702 103464-704
5-Methylsalicylaldehyde 99%
5-Methylsalicylaldehyde
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 217 °C (760 mmHg)
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00274297
CAS Number: 613-84-3

Order Now

Specification Test Results

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 53 to 57 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR