5-(Trifluoromethoxy)salicylaldehyde 99%

Warning

Synonyms: 2-Hydroxy-5-(trifluoromethoxy)benzaldehyde

AC39820-0010 AC39820-2500
103464-690EA 80.8 USD
103464-690 103464-688
5-(Trifluoromethoxy)salicylaldehyde 99%
5-(Trifluoromethoxy)salicylaldehyde
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 82 °C (60 mmHg)
Melting Pt: 31…33 °C
Storage Temperature: Ambient
MDL Number: MFCD00075249
CAS Number: 93249-62-8

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 30 to 34 °C
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR