3-Methylsalicylaldehyde 98%

Warning

Synonyms: 2-Hydroxy-3-methylbenzaldehyde

AC39854-0010 AC39854-0050
103464-678EA 32.8 USD
103464-678 103464-680
3-Methylsalicylaldehyde 98%
3-Methylsalicylaldehyde
Formula: C₈H₈O₂
MW: 136.1479 g/mol
Boiling Pt: 203…205 °C (730 mmHg)
Density: 1.134 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00040798
CAS Number: 824-42-0

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Liquid
Infrared spectrum Conforms
Refractive index 1.5620 to 1.5650 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR