2-Hexanone 98%

Danger

Synonyms: Methyl butyl ketoneButyl methyl ketone

AC14688-1000 AC14688-0025 AC14688-5000
103525-630EA 259 USD
103525-630 103525-626 103525-628
2-Hexanone 98%
2-Hexanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 127 °C (760 mmHg)
Melting Pt: –57 °C
Density: 0.81 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009482
CAS Number: 591-78-6
EINECS: 209-731-1
UN: 1224
ADR: 3,III
Merck Index: 12,06112

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR