2-Fluoro-6-methoxybenzoic acid 97%

Warning

AC43636-0010 AC43636-0050
103464-648EA 30.2 USD
103464-648 103464-650
2-Fluoro-6-methoxybenzoic acid 97%
2-Fluoro-6-methoxybenzoic acid
Formula: C₈H₇FO₃
Melting Pt: 85…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00671764
CAS Number: 137654-21-8

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 85 to 90 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR