2'-Fluoro-4'-methoxyacetophenone 99%

Warning

AC26354-0250 AC26354-1000 AC26354-0050
103464-614EA 40.8 USD
103464-614 103464-618 103464-616
2'-Fluoro-4'-methoxyacetophenone 99%
2'-Fluoro-4'-methoxyacetophenone
Formula: C₉H₉FO₂
MW: 168.17 g/mol
Boiling Pt: 80…85 °C (.1 mmHg)
Melting Pt: 50…54 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00042290
CAS Number: 74457-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR