2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%

Warning

Synonyms: 2-Fluoro-3-(trifluoromethyl)benzeneboronic acid

AC43041-0010
103515-268EA 55.6 USD
103515-268
2-Fluoro-3-(trifluoromethyl)phenylboronic acid 97%
2-Fluoro-3-(trifluoromethyl)phenylboronic acid
Formula: C₇H₅BF₄O₂
MW: 207.92 g/mol
Melting Pt: 60.9…62.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07784336
CAS Number: 157834-21-4

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 64 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR