2-Ethoxy-3-pyridineboronic acid pinacol ester 95%

Warning

Synonyms: 2-Ethoxy-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-pyridine2-Ethoxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine2-Ethoxypyridine-3-boronic acid pinacol ester

AC45436-2500 AC45436-0010 AC45436-0050
103463-518EA 942 USD
103463-518 103463-516 103463-520
2-Ethoxy-3-pyridineboronic acid pinacol ester 95%
2-Ethoxy-3-pyridineboronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
MW: 249.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07366721
CAS Number: 848243-23-2

Order Now

Specification Test Results

Appearance Yellow Viscous liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR