(2-Bromoethyl)cyclohexane 98%

Warning

Synonyms: 1-Bromo-2-cyclohexylethane2-Cyclohexylethyl bromide

AC39264-0250 AC39264-0050 AC39264-0010
103464-584EA 20.4 USD
103464-584 103464-588 103464-586
(2-Bromoethyl)cyclohexane 98%
(2-Bromoethyl)cyclohexane
Formula: C₈H₁₅Br
MW: 191.11 g/mol
Boiling Pt: 209 °C (760 mmHg)
Melting Pt: –57 °C
Density: 1.221 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00019398
CAS Number: 1647-26-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR