2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95%

Danger

Synonyms: 3-((Bis(diisopropylamino)phosphino)oxy)propanenitrileBis(diisopropylamino)(2-cyanoethoxy)phosphine

AC37974-0010 AC37974-0250 AC37974-0050
103528-118EA 64.9 USD
103528-118 103528-122 103528-120
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite 95%
2-Cyanoethyl-N,N,N',N'-tetraisopropylphosphordiamidite
Formula: C₁₅H₃₂N₃OP
MW: 301.41 g/mol
Boiling Pt: 98…100 °C (5 mmHg)
Density: 0.949 g/cm³
Flash Pt: 60 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012213
CAS Number: 102691-36-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear to hazy colorless to yellow Liquid
Infrared spectrum Conforms
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR