2-Chlorothiazole-4-carboxylic acid 97%

Warning

AC42955-0050 AC42955-0010
103464-574EA 202 USD
103464-574 103464-572
2-Chlorothiazole-4-carboxylic acid 97%
2-Chlorothiazole-4-carboxylic acid
Formula: C₄H₂ClNO₂S
MW: 163.58 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD09870033
CAS Number: 5198-87-8

Order Now

Specification Test Results

Appearance White to yellow to orange Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR