2-Chloro-4-pyridineboronic acid pinacol ester 97%

Warning

Synonyms: 2-(2-Chloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane2-Chloropyridine-4-boronic acid Pinacol ester2-Chloropyridine-4-boronic acid pinacol ester2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

AC45422-0050 AC45422-0010
103464-558EA 946.6 USD
103464-558 103464-556
2-Chloro-4-pyridineboronic acid pinacol ester 97%
2-Chloro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₅BClNO₂
MW: 239.51 g/mol
Melting Pt: 63.5…66 °C
Storage Temperature: Ambient
MDL Number: MFCD04039870
CAS Number: 458532-84-8

Order Now

Specification Test Results

Appearance Colorless to white to yellow Solid
Infrared spectrum Conforms
Melting point 62 to 68 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR