2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%

Warning

Synonyms: 2-Chloro-2',6'-dimethylacetanilideN-(2,6-Dimethylphenyl)chloroacetamideN-(2,6-Dichlorophenyl)-2-chloroacetamide

AC13544-0100
103464-540EA 67 USD
103464-540
2-Chloro-N-(2,6-dimethylphenyl)acetamide 98%
2-Chloro-N-(2,6-dimethylphenyl)acetamide
Formula: C₁₀H₁₂ClNO
MW: 197.66 g/mol
Melting Pt: 143…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00000926
CAS Number: 1131-01-7
EINECS: 214-460-7

Order Now

Specification Test Results

Appearance White to brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 143 to 150 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR