2-Chloro-6-methyl-3-(trifluoromethyl)pyridine 97%

Warning

AC45244-0010 AC45244-0050
103464-516EA 254.7 USD
103464-516 103464-518
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine 97%
2-Chloro-6-methyl-3-(trifluoromethyl)pyridine
Formula: C₇H₅ClF₃N
Boiling Pt: 84…86 °C (24Torr)
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD11226678
CAS Number: 1099597-74-6

Order Now

Specification Test Results

Appearance White to yellow to orange Low melting solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR